TCR beta Monoclonal Antibody (H57-597), eFluor™ 506, eBioscience™ - FAQs
TCR beta Monoclonal Antibody (H57-597), eFluor™ 506, eBioscience™ - FAQs
View additional product information for TCR beta Monoclonal Antibody (H57-597), eFluor™ 506, eBioscience™ - FAQs (69-5961-80, 69-5961-82)