IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 506, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 506, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio17B7), eFluor™ 506, eBioscience™ - FAQs (69-7177-80, 69-7177-82)