IL-17A Monoclonal Antibody (eBio64DEC17), eFluor™ 506, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio64DEC17), eFluor™ 506, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio64DEC17), eFluor™ 506, eBioscience™ - FAQs (69-7179-42, 69-7179-41)