IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 506, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 506, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 506, eBioscience™ - FAQs (69-7319-41, 69-7319-42)