CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 506, eBioscience™ - FAQs
CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 506, eBioscience™ - FAQs (69-9985-82, 69-9985-80)