Nalgene™ Large Polypropylene Sterilizing Pans
Thermo Scientific™

Nalgene™ Large Polypropylene Sterilizing Pans

Dry and sterilize instruments and labware with these large, polypropylene sterilizing pans, featuring rolled edges for extra stiffness and molded-in handles for easy carrying.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Unit Size
6900-0010PK
also known as 6900-0010
9.5 LEach
6900-00109.5 LCase of 6
6900-002020 LCase of 6
6900-0020PK
also known as 6900-0020
20 LEach
Catalog number 6900-0010PK
also known as 6900-0010
Price (USD)
63.34
Online exclusive
79.20
Save 15.86
Each
Capacity (Metric):
9.5 L
Unit Size:
Each
Price (USD)
63.34
Online exclusive
79.20
Save 15.86
Each

Dry and sterilize instruments and labware with Thermo Scientific™ Nalgene™ Large Polypropylene Sterilizing Pans. These autoclavable pans are also useful for collecting and washing soiled lab equipment. Pans feature rolled edges for extra stiffness and molded-in handles for easy carrying.

  • Pans are tapered for nesting to save space
  • Heavy wall construction will withstand rugged use and repeated autoclaving

Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
DescriptionSterilizing pans are useful for drying and sterilizing instruments, labware; collecting and washing soiled lab equipment
Dimensions (L x W x H)14.25 x 12.25 x 5.675 in.
Height (English)5.675 in.
Length (English)14.25 in.
No. per Case6
No. per Pack1
ShapeRectangular
Width (English)12.25 in.
AutoclavableAutoclavable
Capacity (Metric)9.5 L
Height (Metric)143 mm
Length (Metric)361 mm
MaterialPolypropylene
TypeSterilizing Pan
Width (Metric)311 mm
Unit SizeEach

Frequently asked questions (FAQs)

Do you offer any trays or pans for secondary containment of reagent bottles?

Yes, 6900 series polypropylene or 7120 series high-density polyethylene trays can be used for this purpose.