Nalgene™ Large Polypropylene Sterilizing Pans
Thermo Scientific™

Nalgene™ Large Polypropylene Sterilizing Pans

Dry and sterilize instruments and labware with these large, polypropylene sterilizing pans, featuring rolled edges for extra stiffness and molded-in handles for easy carrying.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Unit Size
6900-002020 LCase of 6
6900-00109.5 LCase of 6
6900-0010PK
also known as 6900-0010
9.5 LEach
6900-0020PK
also known as 6900-0020
20 LEach
Catalog number 6900-0020
Price (USD)
429.10
Online exclusive
644.00
Save 214.90
Case of 6
Capacity (Metric):
20 L
Unit Size:
Case of 6
Price (USD)
429.10
Online exclusive
644.00
Save 214.90
Case of 6

Dry and sterilize instruments and labware with Thermo Scientific™ Nalgene™ Large Polypropylene Sterilizing Pans. These autoclavable pans are also useful for collecting and washing soiled lab equipment. Pans feature rolled edges for extra stiffness and molded-in handles for easy carrying.

  • Pans are tapered for nesting to save space
  • Heavy wall construction will withstand rugged use and repeated autoclaving

Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
DescriptionSterilizing pans are useful for drying and sterilizing instruments, labware; collecting and washing soiled lab equipment
Dimensions (L x W x H)21.375 x 17.125 x 5.125 in.
Height (English)5.125 in.
Length (English)21.375 in.
No. per Case6
No. per Pack1
ShapeRectangular
Width (English)17.125 in.
AutoclavableAutoclavable
Capacity (Metric)20 L
Height (Metric)130 mm
Length (Metric)543 mm
MaterialPolypropylene
TypeSterilizing Pan
Width (Metric)435 mm
Unit SizeCase of 6

Frequently asked questions (FAQs)

Do you offer any trays or pans for secondary containment of reagent bottles?

Yes, 6900 series polypropylene or 7120 series high-density polyethylene trays can be used for this purpose.