White 96-Well Immuno Plates, Immulon, Round-bottom, Microfluor 1, 230μL - FAQs
White 96-Well Immuno Plates, Immulon, Round-bottom, Microfluor 1, 230μL - FAQs
View additional product information for White 96-Well Immuno Plates - FAQs (7572, 7571, 6905TS, 436111, 9502887, 436110, 7705, 7905)
2 product FAQs found
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.
Why would I use a black or white (instead of clear) immuno plate?
Black plates are often used in assays that make use of a fluorescent reporter because the opaque black surface effectively isolates the signal from each well in order to eliminate “crosstalk” or signal bleed. White plates also isolate well-to-well signals somewhat, but also provide a reflective background to direct luminescent output to the reader, and so often serve as a better choice to improve dynamic range from a luminescent reporter.