Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Thermo Scientific™

Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover

Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.
Have Questions?
Catalog NumberDimensions (L x W x H)
6910-0618PK
also known as 6910-0618
18 x 6 x 2.6 in.
Catalog number 6910-0618PK
also known as 6910-0618
Price (USD)
97.02
Online exclusive
121.30
Save 24.28
Each
Dimensions (L x W x H):
18 x 6 x 2.6 in.
Price (USD)
97.02
Online exclusive
121.30
Save 24.28
Each

Reduce breakage and chipping of glass pipette tips with the Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray. Horizontal design allows pipettes to be fully immersed in disinfectant. Rigid plastic tray is stackable, lightweight, quiet and less expensive than stainless steel. Cover fits into groove of pan to minimize spillage of hazardous materials.

  • Holds pipets up to 40cm long in a stackable, lightweight alternative to stainless steel
  • Can also be used to sterilize small labware and instruments—resists aggressive disinfectants
  • Has groove for cover to minimize spillage of hazardous materials and a hand grip on the cover
  • Autoclavable

Compliance:

  • Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens.
  • Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of pipets are required before cleaning and disposal

Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
Certifications/ComplianceMeets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens. Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of
DescriptionSterilizing tray with rigid plastic construction
Dimensions (L x W x H)18 x 6 x 2.6 in.
For Use WithHolds pipets up to 40cm long
Height (English)2.6 in.
Length (English)18 in.
No. per Case6
No. per Pack1
ShapeRectangular
Width (English)6 in.
AutoclavableAutoclavable
Height (Metric)6.7 cm
Length (Metric)46 cm
MaterialPolypropylene
TypeSterilizing Tray
Width (Metric)15 cm
Unit SizeEach