Slide-A-Lyzer™ MINI Dialysis Devices, 10K MWCO
Thermo Scientific™

Slide-A-Lyzer™ MINI Dialysis Devices, 10K MWCO

Perform sample buffer exchange with these 10,000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
69574Dialysis Unit, Microcentrifuge Tubes1 Kit (10 Devices)
69570Dialysis Unit50 Devices
69572
also known as 000000
Dialysis Unit250 Devices
69576Dialysis Unit, Float Buoy1 Kit (10 Devices)
88401Dialysis Unit, 15 mL Conical Tube25 Devices
88404Dialysis Unit, 50 mL Conical Tube25 Devices
Catalog number 69574
Price (USD)
99.65
Online exclusive
110.00
Save 10.35
Each
Product Type:
Dialysis Unit, Microcentrifuge Tubes
Quantity:
1 Kit (10 Devices)
Request bulk or custom format
Price (USD)
99.65
Online exclusive
110.00
Save 10.35
Each
Slide-A-Lyzer MINI Dialysis Devices, 10K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules greater than 10 kDa in volumes from 10 μL to 2 mL.
Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

Features of Slide-A-Lyzer MINI Dialysis Devices, 10K MWCO:
• Three convenient sizes—dialysis devices for 0.1, 0.5, and 2 mL samples fit standard 1.5-mL micro-centrifuge, 15- and 50-mL conical tubes, respectively
• Excellent sample recoveries—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
• One-step protocol—pipette sample into the MINI device and place in tube containing the dialysis buffer; no laborious assembly, device preparation, or expensive equipment are required
• 100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
• Time savings—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

These 10K MWCO dialysis devices are available in three sizes (0.1, 0.5, and 2 mL capacities), and are designed to allow easy sample addition and removal using a standard laboratory pipette. The MINI devices are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4–8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with a number of common chemicals and buffers.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker. The 0.5- and 2-mL MINI dialysis devices fit in and are capped by inserting them into the included 15- and 50-mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI dialysis devices, low molecular weight contaminant removal and buffer exchange and desalting can be accomplished within 4–8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

Using the 0.1 mL MINI device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

For Research Use Only. Not for use in diagnostic procedures.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

The 0.5mL and 2mL MINI Dialysis Devices fit in and are capped by inserting them into the included 15mL and 50mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI Dialysis Devices, low molecular weight contaminant removal, buffer exchange and desalting, can be accomplished within 4 to 8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

Specifications
Kit ContentsDialysis Units, Microcentrifuge Tubes
MembraneCellulose Membrane, Dialysis
Product LineSlide-A-Lyzer
Product TypeDialysis Unit, Microcentrifuge Tubes
Purification TargetBuffer Exchange, Protein
Quantity1 Kit (10 Devices)
FormatMINI Dialysis Device
MWCO10 kDa
Sample Volume (Metric)0.1 mL
Unit SizeEach
Contents & Storage
Store at room temperature

Frequently asked questions (FAQs)

With what chemicals is the Slide-A-Lyzer MINI Dialysis Unit compatible?

The Slide-A-Lyzer MINI Dialysis Unit is compatible with alcohols, ketones, esters, oxides, and solvents containing nitrogen.
Alkalis: 1N sodium hydroxide or 1N ammonium hydroxide may cause pore swelling or shrinkage; not recommended to be used with 1N potassium hydroxide
Acids: Will survive weak acids (100% acetic or formic acids), or ~25% phosphoric or sulfuric acids; it should not be used with >25% strong acids, although a weak solution of a strong acid may work

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What percent of my sample should I expect to lose during dialysis when using the Slide-A-Lyzer MINI Dialysis Unit?

In general, polypropylene and regenerated cellulose have very low nonspecific binding properties. The percent recovery of a sample will depend on the unique properties of the sample and on the initial concentration of the sample.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What materials are used to manufacture the Slide-A-Lyzer MINI Dialysis Unit?

The membrane is regenerated cellulose and the plastic is a polypropylene copolymer. Both materials have very low protein and nucleic acid binding properties.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How long does it take to dialyze with the Slide-A-Lyzer MINI Dialysis Unit?

The Slide-A-Lyzer MINI Dialysis Unit can convert 100 µL of pH 2.8 buffer to pH 9.4 dialyzing against 1 liter bicarbonate buffer, pH 9.4 in less than 10 minutes.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How do I use the Slide-A-Lyzer MINI Dialysis Unit?

- Wearing gloves, apply sample (0.01-0.1 mL) using a pipette.
- OPTIONAL: Soak the Slide-A-Lyzer MINI Dialysis Unit to remove trace contaminants.
- Cap the Slide-A-Lyzer MINI Dialysis Unit (to minimize loss) and snap it into the float (sold separately).
- Insert the float into the beaker containing the dialysate.
- Recover sample.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.