SureSTART™ Red Polypropylene 9 mm AVCS™ Screw Caps with Red Silicone/White PTFE Septa, 1000/pack, Level 2 - FAQs
SureSTART™ Red Polypropylene 9 mm AVCS™ Screw Caps with Red Silicone/White PTFE Septa, 1000/pack, Level 2 - FAQs
View additional product information for SureSTART™ 9 mm Screw Caps, Level 2 High-throughput Applications - FAQs (6ASC9STBS1F, 6ASC9ST1G, 6ASC9ST1F, 6ASC9ST1B, 6ASC9ST1, 6ASC9STB1, 6ASC9RT1, 6ASC9STBS1T, 6ASC9RT1Y, 6ASC9STB1T, 6ASC9RT1T, 6ASC9RT1R, 6ASC9STBS1, 6ASC9STB1F, 6ASC9ST1RT, 6ASC9RT1G, 6ASC9RT1F, 6ASC9RT1RF, 6ASC9CTRRT1, 6ASC9RT1B, 6ASC9ST1Y, 6ASC9ST1RF, 6ASC9SPI1, 6ASC9RT1RT, 6ASC9ST1T, 6ASC9ST1R)
1 product FAQs found
What are AVCS caps?
Thermo Scientific Advanced Vial Closure System (AVCS) caps are polypropylene caps that are compatible with all 9 mm short thread screw vials, although we recommend that they are used with our Thermo Scientific SureStop vials to get maximum benefit of the two technologies. There is a leveling feature within the AVCS designed caps that provide optimal liquid sealing. The innovative features found on the AVCS cap also give more autosampler compatibility with less autosampler interruptions, as a result of misaligned cap tilting, dislodged septa, or septa push through.
AVCS caps not only have the most effective seal but they also contain significantly less leachables than bonded caps. You'll see a noticeably cleaner spectra when you use SureStop and AVCS as an alternative to bonded caps. The AVCS caps also come with a range of different septa materials.