SureSTART™ 0.3 mL Polyspring Plastic Inserts (Level 1) for 2 mL Vials, Conical with Polyspring, 5000/pack - FAQs
SureSTART™ 0.3 mL Polyspring Plastic Inserts (Level 1) for 2 mL Vials, Conical with Polyspring, 5000/pack - FAQs
View additional product information for SureSTART™ Polypropylene Polyspring Inserts for 2 mL HPLC & GC Vials, Level 1 - FAQs (6EME03CPPSP, 6EME03CPPSPT, 6EME03CPPSPF)
1 product FAQs found
What are SureSTART micro inserts?
The micro inserts have a small total volume capacity of <400 µL and can be classified as either Polyspring, conical, or flat-bottom within the SureSTART Collection. Micro inserts are placed and sealed inside a vial so they can be used in conventional autosampler racks. The smaller 6 mm diameter of the insert gives better sample recovery than a routine flat based vial with conical inserts being most effective with residual volumes of 1-2 µL. Many are held in place by a Polyspring, which cushions and centers the insert.