Specifications
AutoclavableNot Autoclavable
Base StyleConical with Polyspring
Certifications/ComplianceSpecification Certified
Diameter (Metric) Inner6 mm
DisposableSingle-use
For Use With (Application)Micro Sampling
For Use With (Equipment)2 mL Vials
Height (Metric)30 mm
Insert TypeInserts with spring for HPLC and GC Vials
Packaging Type10 Packs of 100
Product TypePolyspring Insert
Volume (Metric) Residual2 μL
Volume (Metric) Total0.325 mL
Volume (Metric) Usable0.25 mL
ColorClear
Diameter (Metric) Outer6 mm
MaterialPolypropylene
Product LineSureSTART Level 1
Quantity1000
SterilityNon-sterile
Unit SizeCase of 1000