Search
Search

| Catalog Number | Product Type | Thread Style | Quantity |
|---|---|---|---|
| 6PCV11G | Crimp | 11 mm | 100/Pk. |
| 6PSV9-1PG | SureStop Screw Thread | 9 mm SureStop Short Thread | 100/Pk. |
| 6PSV9-1PGF | SureStop Screw Thread | 9 mm SureStop Short Thread | 5000/Pk. |
| 6PSV9-1PGT | SureStop Screw Thread | 9 mm SureStop Short Thread | 1000/Pk. |
Thermo Scientific™ SureSTART™ 2 mL GOLD-Grade Glass Screw and Crimp Top Vials (Level 3) are the elite choice for high-sensitivity LC-MS, GC-MS, and UHPLC workflows where analytes such as polar compounds or those affected by pH changes demand maximum sample stability and ultra-low adsorption.
These vials are made from Thermo Scientific™ GOLD-Grade low-adsorption glass, offering:
The screw-top version features Thermo Scientific™ SureStop™ technology, which reduces cap tilt, septum push-through, and evaporation – maintaining sample integrity throughout demanding analyses. To ensure optimal performance, pair with Thermo Scientific™ SureSTART™ Advanced Vial Closure System (AVCS™) 9 mm caps and septa.
The crimp-top version, combine with 11 mm Thermo Scientific™ SureSTART™ crimp caps and septa. This will provide a tight, solvent-resistant seal ideal for volatile samples and long-run LC/GC applications, ensuring maximum sample security under challenging conditions.
Key Features
Performance Level 3 – GOLD-Grade for Ultra-Trace Applications
Specification Certified
Lot Traceable