SureSTART™ MSCERT 2 mL Short Thread Screw Amber Glass SureStop™ Vial and 9 mm AVCS™ Screw Cap Kits with Pre-slit Blue Silicone/Clear PTFE Septa, Level 3 - FAQs
SureSTART™ MSCERT 2 mL Short Thread Screw Amber Glass SureStop™ Vial and 9 mm AVCS™ Screw Cap Kits with Pre-slit Blue Silicone/Clear PTFE Septa, Level 3 - FAQs
View additional product information for SureSTART™ MSCERT Screw Vial and Cap Kits, Level 3 High Performance Applications - FAQs (6PMSCK540W, 6PMSCK651W, 6PMSCK541W, 6PMSCK641W, 6PMSCK535W, 6PMSCK611W, 6PMSCK534W, 6PMSCK601W)
6 product FAQs found
Are SureSTOP vials compatible with standard autosamplers, or do I need special equipment?
SureSTOP vials are designed for compatibility with nearly every LC or GC autosampler. They are part of the SureSTART family, which is tested for autosampler compatibility and specification certified for physical dimensions.
How do SureSTOP vials help with evaporation control compared to generic vials?
Under stress conditions (e.g., 1.3 mL methanol at 40 °C for 72 h), SureSTOP vials with AVCS closures lose significantly less solvent compared with generic screw-top or even crimp vials. The “definite stop” means the cap is tightened correctly (neither under nor over-tightened) so septum compression is predictable, and cap-tilt or septum displacement (which lead to evaporation paths) are minimized.
In workflows where solvent loss or sample concentration change is critical (as for very low volumes or volatile analytes), SureSTOP vials offer measurable improvement.
What types of applications are SureSTOP vials well-suited for?
SureSTOP vials are well-suited for:
• GC, LC and MS workflows where volatile or semi-volatile solvents are used and evaporation or sample loss is a concern.
• High-sensitivity or trace-analysis workflows where variation in sealing can impact reproducibility.
• Labs where autosampler failures (due to cap/tilt/septum issues) are affecting throughput.
How are SureSTOP vials different from standard screw-top autosampler vials?
Key aspects of SureSTOP vials include:
• The “definite stop” design prevents over-tightening (which might deform the septum or the vial neck) and under-tightening (leading to leaks or evaporation).
• Achieve the best performance with caps that have Advanced Vial Closure System (AVCS) technology that levels and aligns the cap properly, reducing cap tilt and septum displacement.
• Demonstrated lower solvent loss (evaporation) in stressed tests compared with generic vial and closure combinations.
What problems do SureSTOP vials help solve?
The SureSTOP vials are 2 mL screw-thread (9 mm) vials that incorporate a patented “definite stop point” in the vial finish (alignment ring) so when you use them with AVCS (Advanced Vial Closure System) 9 mm screw caps, you cannot under- or over-tighten the closure. This feature helps solve problems such as inconsistent sealing, variable septum compression, vial/septa push through, sample evaporation, or autosampler failures due to poorly sealed vials.
What are SureStop vials?
In addition to the features on standard 2 mL vials, SureStop vials also have a patented definite stop point (alignment ring) in the design of the vial finish. This prevents the vial from being an optimal seal, which eliminates under- and over-sealing by the cap/closure and user-to-user variance.
SureStop vials are designed for use with Advanced Vial Closure System (AVCS) screw caps. They improve transfer reliability and give a more secure liquid seal than standard screw vials. SureStop vials are available as a 2 mL as either clear or amber options and are highly recommended for use with GC and LC applications.