Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Actual product may vary
Thermo Scientific™

Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor

Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or Material
700001TS
also known as 700001
Low Ionic Strength pH Test Kit
700402pH Buffer B
700003pHISA Low Ionic Strength Adjustor
700702pH Buffer A
700902TS
also known as 700902
pH Buffer C
Catalog number 700001TS
also known as 700001
Price (USD)
375.30
Online exclusive
417.00
Save 41.70
Each
Chemical Name or Material:
Low Ionic Strength pH Test Kit
Price (USD)
375.30
Online exclusive
417.00
Save 41.70
Each

Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.

Features:

  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample

Pure Water pH Test Kit:

  • Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
  • For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certificate of AnalysisYes
Certifications/ComplianceISO 9001
Chemical Name or MaterialLow Ionic Strength pH Test Kit
PackagingKit
Traceability to NISTYes
For Use With (Application)Provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water.
For Use With (Equipment)pH Meter and Electrode
GradeCertified
Physical FormLiquid
Quantity4 x 475 mL pH 6.97 Buffer, 2 x 475 mL pH 4.10 Buffer, 2 x 60 mL pHISA Adjuster
Solution TypepH Buffer
pH6.97
Unit SizeEach