Orion™ Total Alkalinity Test Kit
Thermo Scientific™

Orion™ Total Alkalinity Test Kit

Save time by eliminating additional equipment set-up, titrations and calculations with this total alkalinity test kit that provides two-step direct measurement.
Have Questions?
Change viewbuttonViewtableView
Quantity:
475 mL
4 x 475 mL
Type:
View all
Total Alkalinity Test Kit
Alkalinity Standard/Control Refill
Catalog number 700010TS
also known as 700010
Price (USD)
342.00
Each
Quantity:
475 mL
Type:
Total Alkalinity Test Kit
Request bulk or custom format
Price (USD)
342.00
Each
Specifications
ColorClear
CompositionNon Hazardous Media
For Use With (Application)two-step direct measurement of total alkalinity and pH using a pH electrode; Saves time by eliminating additional equipment set-up, tedious titrations and calculations
For Use With (Equipment)pH Electrode
FormatLiquid
View more

Save time by eliminating additional equipment set-up, titrations and calculations. The Thermo Scientific™ Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH using a pH electrode.

  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity

Includes:

  • One 1 pint (475mL) bottle alkalinity reagent (700011)
  • One 1 pint (475mL) bottle alkalinity standard/control (700012)
  • Total alkalinity conversion chart
  • Application procedure No. 517, Total Alkalinity in Natural Waters

Note:

The following part number has been changed due to a recent system upgrade.

Old Part Number: 700011

New Part Number: 700011-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

Warranty: Out of box

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.