RNASE AWAY Squeeze Bottle 250mL (8oz) - FAQs
RNASE AWAY Squeeze Bottle 250mL (8oz) - FAQs
View additional product information for RNASE AWAY Squeeze Bottle 250mL (8oz) - FAQs (7000TS1)
1 product FAQs found
Do you have a Technical data sheet (TDS) for RNase AWAY Surface Decontaminant?
We have a safety data sheet (SDS) and COA for RNase AWAY Surface Decontaminant. Visit our Documents and Certificates page to search for and request these documents.