NanoDrop™ Eight On-site Installation & Recertification
Actual product may vary
Thermo Scientific™
NanoDrop™ Eight On-site Installation & Recertification
Rely on our trained experts to perform on-site installation or recertification of your Thermo Scientific™ NanoDrop™ Eight. Services include on-site installation of NanoDrop Eight PC Software and 21 CFR Part 11 compliance software, basic user orientation, instrument installation qualification and operational qualification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
701-059049ND Eight OQ Recertification
701-059048ND Eight Validated Install
701-059050ND Eight Dig Signature SW OQ
Catalog number 701-059049
Price (USD)
-
Type:
ND Eight OQ Recertification
Rely on our trained experts to perform on-site installation or recertification of your Thermo Scientific™ NanoDrop™ Eight. Services include on-site installation of NanoDrop Eight PC Software and 21 CFR Part 11 compliance software, basic user orientation, instrument installation qualification and operational qualification. All engineer labor and travel charges for installation within continental US. Additional charges may apply for outside of continental US or remote Canadian regions. Separate purchase of 21 CFR Part 11 compliance software for NanoDrop Eight Spectrophotometers and NanoDrop Eight IQ/OQ Kit required.
NanoDrop Eight Validated Installation
Features:
On-site installation of instrument
On-site installation of NanoDrop Eight and SciVault software
Basic user orientation
Instrument installation Qualification and Operational Qualification
All engineer labor and travel charges for installation within continental US. Additional charges may apply for outside of continental US or remote Canadian regions.
Requires:
Separate purchase of SciVault for NanoDrop Eight
Separately purchased NanoDrop Eight IQ/OQ Kit
Exclusions:
Consumables such as cuvettes, pipettes, flash drives, printer paper
NanoDrop Eight Digital Signature Software OQ
Features:
Execution of NanoDrop Eight SciVault OQ
Requires:
Separate purchase of SciVault for NanoDrop Eight
Requires the purchase of 701-059048 NanoDrop Eight Validated Installation
NanoDrop Eight OQ Recertification
Annual Operational Qualification
All engineer labor and travel charges for OQ within continental US. Additional charges may apply for outside of continental US or remote Canadian regions.
Requires:
Separate purchase of SciVault for NanoDrop Eight
Separately purchased NanoDrop Eight IQ/OQ Kit or NanoDrop Eight Requalification Kit
Exclusions:
Consumables such as cuvettes, pipettes, flash drives, printer paper
Specifications
For Use With (Equipment)NanoDrop Eight Spectrophotometers
IncludesAnnual Operational Qualification, All engineer labor and travel charges for installation within continental US. Additional charges may apply for outside of continental US or remote Canadian regions. Requires: Separate purchase of 21 CFR Part 11 compliance software for NanoDrop Eight Spectrophotometers and NanoDrop Eight IQ/OQ Kit or NanoDrop Eight Requalification Kit
TypeND Eight OQ Recertification
Unit SizeEach
Frequently asked questions (FAQs)
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.