You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Chromatography Columns and Cartridges
▸
HPLC Columns and Cartridges
▸
HPLC Columns
▸
BetaSil™ C6 HPLC Columns, 5μm Particle Size
Actual product may vary
BetaSil™ C6 HPLC Columns, 5μm Particle Size
High surface area with high bonded phase coverage
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
70305-254630
250 mm
4.6 mm
70305-154630
150 mm
4.6 mm
70305-154030
150 mm
4 mm
70305-124630
125 mm
4.6 mm
4 Options
Catalog number
70305-254630
Price (USD)
1,334.70
Each
Add to cart
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Price (USD)
1,334.70
Each
Add to cart
High surface area with high bonded phase coverage
Thermo SCientific chemistry on 100&Angst; silica
Specifications
Column Format
Analytical Column
Column Type
Reversed Phase
Endcapped
Yes
Packing Material
Silica
Stationary Phase
C6
USP Type
L15
Carbon Load
11%
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Particle Size
5 μm
Phase
Reversed Phase
Pore Size
100 Å
Product Line
Beta
Unit Size
Each