Product Image
Applied Biosystems™

Dynabeads™ EPEC/VTEC O26

For the rapid selective concentration of E.coli 026. Method can be performed manually or can be automated by using aMás información
Have Questions?
Número de catálogoCantidad
710132 mL
Número de catálogo 71013
Precio (MXN)
-
Cantidad:
2 mL
For the rapid selective concentration of E.coli 026. Method can be performed manually or can be automated by using a Thermo Scientific KingFisher mL Food Protection Purification System.

This Product Contains: Dynabeads coated with anti-E.coli O26 antibodies

Sensitivity: Sensitivity is very high and enables detection of 1 viable organism in 25g of sample or as few as 100 organisms / ml after pre enrichment
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Para utilizar con (equipo)Dynabeads™ MPC™, KingFisher mL Food Protection Purification System
Línea de productosDynabeads
Condiciones de envíoRoom Temperature
Cantidad2 mL
Especies dianaE. coli
Unit SizeEach
Contenido y almacenamiento
Store in refrigerator (2–8°C).

Preguntas frecuentes

What are the advantages of the Dynabeads magnetic beads immunomagnetic separation (IMS) system?

High sensitivity--reduces false negative results
High recovery--greater than 60% and as high as 90%
Read only one clean slide well--improves workflow and accuracy
Antibody based selection system--provides increased specificity

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the benefits of the Dynabeads magnetic beads microbiology enrichment products?

Dynabeads magnetic beads microbiology enrichment products offer the following advantages:

Simple protocol
Cost effective
Shelf-stable reagents
Easy QC
High sensitivity
Saves time (for Salmonella in processed food, the assay is 24 hours faster compared to standard methods because immunomagnetic separation (IMS) replaces conventional selective enrichment; also saves time in confirmation procedure because you already have the colony isolated)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.