CaptureSelect™ Biotin Anti-FIX Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-FIX Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-FIX Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and plasma derived human Factor IX (FIX).
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
7103002500500 μg
7103002100100 μg
카탈로그 번호 7103002500
제품 가격(KRW)
-
수량:
500 μg

CaptureSelect Biotin Anti-FIX Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and plasma derived human Factor IX (FIX). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-FIX Conjugate allows you to:

  • Detect, quantitate, and characterize recombinant and plasma derived human FIX

Applications for CaptureSelect Biotin Anti-FIX Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
사양
콘주게이트Biotin and Other Haptens
제품라인CaptureSelect
제품 유형Biotin Anti-FIX Conjugate
수량500 μg
Unit SizeEach
구성 및 보관
Store in freezer (-5°C to -30°C).