CaptureSelect™ Biotin Anti-Factor X Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-Factor X Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-Factor X Conjugate consists of a 13-kDa recombinant single domain anti-Factor X antibody fragment chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the antibody fragment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103702500500 μg
7103702100100 μg
Catalog number 7103702500
Price (CLP)
-
Quantity:
500 μg

CaptureSelect Biotin Anti-Factor X Conjugate consists of a 13-kDa recombinant single domain anti-Factor X antibody fragment chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the antibody fragment when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize the human coagulation Factor X and Factor Xa.

The antibody fragment specifically binds to human Factor X and its activated form Factor Xa from plasma and recombinant sources. It is highly selective for human Factor X and Xa with no affinity for other human plasma proteins. The ligand binds in a calcium dependent manner, so full release of the bound Factor X or Factor Xa molecule is achieved by addition of chelating agents like EDTA. Suitable binding and target-release conditions are listed below:

  • Binding condition: 50 mM Tris, 150 mM NaCl, 5 mM CaCl2 pH 7
  • Release condition: 50 mM Tris, 150 mM NaCl, 5 mM EDTA pH 7

Applications for CaptureSelect Biotin Anti-Factor X Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with a molecular weight of approximately 59 kDa. While Factor X normally circulates in the plasma as an inactive molecule, the activation of Factor X is involved in both the intrinsic and extrinsic coagulation pathways.

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeBiotin & Analogs
Product LineCaptureSelect
Product TypeConjugate
Shipping ConditionDry Ice
Quantity500 μg
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).