SSTR2 Recombinant Superclonal™ Antibody (11HCLC) - FAQs
SSTR2 Recombinant Superclonal™ Antibody (11HCLC) - FAQs
View additional product information for SSTR2 Recombinant Superclonal™ Antibody (11HCLC) - FAQs (711091)
1 product FAQs found
Do you offer SSTR2 antibodies for western blot detection?
We offer 5 SSTR2 polyclonal antibodies that target human and have been verified for western blot detection (https://www.thermofisher.com/antibody/primary/target/sstr2/filter/proteintarget/SSTR2/application/Western%20Blot/species/Human). Their tested dilutions for western blot detection are listed below.
Catalog Number and related western blot dilution information:
PA3-109    1:250
711091     1-2 µg/mL
PA3-16736   1:100-1:1000
PA5-85431   1:500-1:3000
BS-1138R   Assay Dependent
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.