VersaTREK™ Myco PZA Kit
VersaTREK™ Myco PZA Kit
VersaTREK™ Myco PZA Kit
VersaTREK™ Myco PZA Kit
Thermo Scientific™
VersaTREK™ Myco PZA Kit
Perform accurate, FDA-cleared Mtb susceptibility testing utilizing pyrazinamide (PZA) with the Thermo Scientific™ VersaTREK™ Myco PZA Kit.
Have Questions?
Catalog NumberPackaging TypeProduct Type
7116-7050 Tests/KitMicrobiology Media
Catalog number 7116-70
Price (USD)
-
Packaging Type:
50 Tests/Kit
Product Type:
Microbiology Media
Perform accurate, FDA-cleared Mtb susceptibility testing utilizing pyrazinamide (PZA) with the Thermo Scientific™ VersaTREK™ Myco PZA Kit.
In addition to the VersaTREK Myco Susceptibility Kit (7115-60), the VersaTREK Myco PZA Kit (300 μg/mL) is also available for convenient, simple first line susceptibility testing for Mycobacterium tuberculosis.
The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionVersaTREK Myco PZA Kit
Packaging Type50 Tests/Kit
Quantity2 x 25 mL Pyrazinamide, 4 x 25 mL PZA Rehydration Buffer
Specimen TypeAll Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
Product TypeMicrobiology Media
SpeciesMycobacterium tuberculosis
Target Organism ClassMycobacteria
Test TypeMicrobial Detection, Antimicrobial Susceptibility
Unit SizeEach
Contents & Storage
2°C to 8°C