Nalgene™ HDPE Pans, 10L - FAQs
Nalgene™ HDPE Pans, 10L - FAQs
View additional product information for Nalgene™ HDPE Pans - FAQs (7120-0010, 7120-0010PK, 7120-0020)
1 product FAQs found
Do you offer any trays or pans for secondary containment of reagent bottles?
Yes, 6900 series polypropylene or 7120 series high-density polyethylene trays can be used for this purpose.