Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Thermo Scientific™

Nalgene™ HDPE Pans

Soak labware or form ice baths using these HDPE pans, which are tapered for nesting and chemically resistant to most acids, bases and alcohols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Unit Size
7120-0010PK
also known as 7120-0010
10 LEach
7120-001010 LCase of 6
7120-002020 LCase of 6
Catalog number 7120-0010PK
also known as 7120-0010
Price (USD)
55.28
Online exclusive
61.40
Save 6.12
Each
Capacity (Metric):
10 L
Unit Size:
Each
Price (USD)
55.28
Online exclusive
61.40
Save 6.12
Each

Soak labware or form ice baths using Thermo Scientific™ Nalgene™ HDPE Pans. These pans are tapered for nesting and are chemically resistant to most acids, bases and alcohols.

  • Without covers
  • Rugged and durable, tapered for nesting
  • Molded-in handles and rolled edges for easy carrying and handling
  • Resistant to most acids, bases and alcohols for a multitude of lab uses
.

Note:
These pans are NOT autoclavable.

Specifications
DescriptionRugged, durable pans tapered for nesting
Dimensions (L x W x H)14.125 x 12.125 x 5.625 in. (36 x 31 x 14 cm)
Height (English)5.69 in.
Length (English)14 in.
No. per Case6
No. per Pack1
ShapeRectangular
Width (English)12.3 in.
AutoclavableNot Autoclavable
Capacity (Metric)10 L
Height (Metric)144.5 mm
Length (Metric)356 mm
MaterialHDPE
TypePan
Width (Metric)312 mm
Unit SizeEach

Frequently asked questions (FAQs)

Do you offer any trays or pans for secondary containment of reagent bottles?

Yes, 6900 series polypropylene or 7120 series high-density polyethylene trays can be used for this purpose.