Forma™ 8600 Series -40°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.1L) capacity, 115V/60Hz, 6.5A - FAQs
Forma™ 8600 Series -40°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.1L) capacity, 115V/60Hz, 6.5A - FAQs
View additional product information for Forma™ 8600 Series -40°C Ultra-Low Temperature Chest Freezers - FAQs (717CA)