Thermo Scientific™
Richard-Allan Scientific™ Signature Series Hematoxylin 1 7221
 Related applications:
Anatomical Pathology
Yield visually similar results to Gills 1 and 2 with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 1, a uniquely formulated product.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation
Available in 5-liter packaging
  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
Documents
Manuals & protocols