PCC-54™ Detergent Concentrate, 3 L - FAQs
PCC-54™ Detergent Concentrate, 3 L - FAQs
View additional product information for PCC-54™ Detergent Concentrate - FAQs (72289, 72288)
6 product FAQs found
How can grease and oils be removed using the RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate??
To remove grease and oil, the detergents can be mixed with ethanol (e.g., two parts of a 2% solution to one part ethanol).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What materials are not compatible with the RBS 35 and PCC-54 detergents?
Because these detergents are alkaline, they may damage non-ferrous metals such as aluminum, zinc, magnesium and galvanized iron. Furthermore, neoprene rubber products should not be soaked in these detergents for extended periods of time.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the pH of the RBS 35 and PCC-54 detergents?
The pH of these detergents is slightly greater than 10.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the working concentration of the RBS 35 and PCC-54 detergents?
The working concentration for general-purpose applications is 2% for the concentrates and 0.3% for the automatic washer detergents. A 20% solution can be used for difficult cleaning situations.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the composition of the RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate?
RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate consist of ionic and nonionic surfactants compounded with strong water-softening cleaning powder components.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the difference between RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate?
RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate are similar products and may be used for the same applications. We do not sell RBS 35 Detergent Concentrate in Europe; Europe customers can purchase our PCC-54 35 Detergent Concentrate instead.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.