PCC-Pfree™ Detergent Concentrate - FAQs
PCC-Pfree™ Detergent Concentrate - FAQs
View additional product information for PCC-Pfree™ Detergent Concentrate - FAQs (72290)
1 product FAQs found
Are RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate phosphate-free?
RBS-pF Detergent Concentrate (Cat. Nos. 27959 and 27960), and PCC-Pfree Detergent Concentrate (Cat. No. 72290) are all phosphate-free.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.