BioBasic™ 4 HPLC Columns
Actual product may vary
BioBasic™ 4 HPLC Columns
BioBasic™ 4 HPLC Columns
Thermo Scientific™

BioBasic™ 4 HPLC Columns

Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
72305-101030100 mm1 mm
72305-05103050 mm1 mm
72305-05213050 mm2.1 mm
72305-102130100 mm2.1 mm
72305-104630100 mm4.6 mm
72305-152130150 mm2.1 mm
72305-154630150 mm4.6 mm
72305-252130250 mm2.1 mm
72305-254630250 mm4.6 mm
72305-01210110 mm2.1 mm
72305-01400110 mm4 mm
Catalog number 72305-101030
Price (USD)
1,148.60
Each
Length (Metric):
100 mm
Diameter (Metric):
1 mm
Price (USD)
1,148.60
Each

Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedYes
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Product TypeAnalytical Column
Stationary PhaseC4
Surface Area100 m²/g
Temperature60°C
USP TypeL26
Carbon Load4%
Diameter (Metric)1 mm
Length (Metric)100 mm
Particle Size5 μm
PhaseReversed Phase
Pore Size300 Å
Product LineBioBasic
pH2 to 9
Unit SizeEach