Manostat™ Tygon™ Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Tygon™ Pumping Links
Manostat Tygon pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeDescription
7231025525Simon/Vera, Kate, Compulab RD2
7231017517Simon/Vera, Kate, Compulab RD2
7531014514Preston, Spencer and Compulab RD1
7531025525Preston, Spencer and Compulab RD1
7531016516Preston, Spencer and Compulab RD1
7531017517Preston, Spencer and Compulab RD1
7231013513Simon/Vera, Kate, Compulab RD2
7231016516Simon/Vera, Kate, Compulab RD2
7231014514Simon/Vera, Kate, Compulab RD2
7531013513Preston, Spencer and Compulab RD1
Catalog number 72310255
Price (BRL)
-
Tubing Size:
25
Description:
Simon/Vera, Kate, Compulab RD2
Thermo Scientific™ Manostat Tygon pumping links are for use with Manostat pumps and dispensers.
Specifications
DescriptionSimon/Vera, Kate, Compulab RD2
For Use WithSimon/Vera, Kate and Compulab RD2
Includes5 links/pack
Product LineThermo Scientific
Tubing Size25
Diameter (English) Inner3/16 in.
Diameter (English) Outer0.975 in.
Diameter (Metric) Inner4.8 mm
Diameter (Metric) Outer25 mm
MaterialTygon
Product TypePump Tubing Links
Unit SizeEach