CaptureSelect™ Alexa Fluor™ 647 Anti-AAVX Conjugate
Thermo Scientific™

CaptureSelect™ Alexa Fluor™ 647 Anti-AAVX Conjugate

Thermo Scientific™ CaptureSelect™ Alexa Fluor™ 647 Anti-AAVX Conjugate consists of a 14-kDa recombinant single-domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to adeno-associated virus particles (AAV).
Have Questions?
Catalog NumberQuantity
7233522100100 μg
Catalog number 7233522100
Price (MXN)
-
Quantity:
100 μg

CaptureSelect Alexa Fluor 647 Anti-AAVX Conjugate consists of a 14-kDa recombinant single-domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to adeno-associated virus particles (AAV). The anti-AAVX affinity ligand demonstrates good binding reactivity towards a variety of AAV serotypes, including AAV1 to AAV8, AAVrh10, and synthetic serotypes.

The affinity ligand is chemically conjugated to Alexa Fluor 647 via an appropriate spacer that fully retains the binding reactivity of the ligand, enabling detection of AAV total capsids. Conjugation of the CaptureSelect AAVX affinity ligand with the Alexa Fluor 647 dye combines the high binding affinity of the ligand with the unique characteristics of the dye, which is a bright, far-red-fluorescent dye with excitation for the 594 nm or 633 nm laser lines. This combination makes this product ideally suited for stable signal generation in fluorescent immunoassay (FIA) applications and imaging.

Although anti-AAVX conjugates, including CaptureSelect Biotin Anti-AAVX (Cat. No. 7103522100) and CaptureSelect HRP Anti-AAVX (7303522100), cover a broad range of AAV serotypes, the binding reactivity for AAV9 is minimal and therefore not recommended for use in detection methods for AAV9 capsids. For the detection of AAV9 capsids we recommend using CaptureSelect Biotin Anti-AAV9 (7103332100) and CaptureSelect HRP Anti-AAV9 (7303332100).

Product characteristics

  • Format type: Alexa Fluor, liquid
  • Buffer: PBS
  • Storage: store in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.2 mL
  • Working dilution: the optimal working dilution should be determined empirically for each application

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin Free).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)200 μL
Concentration0.5 mg/mL
For Use With (Application)Detection
Label or DyeAlexa Fluor™ 647
Ligand TypeAAVX Affinity Ligand
Quantity100 μg
TargetMultiple AAV Serotypes
FormLiquid
Product LineCaptureSelect
TypeConjugate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).