Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, holds 240 boxes; 208-230V/60Hz, 12A - FAQs
Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, holds 240 boxes; 208-230V/60Hz, 12A - FAQs
View additional product information for Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (7240D)