Nalgene™ 25mm Syringe Filters, Nylon, 0.2μm pore, Sterile - FAQs
Nalgene™ 25mm Syringe Filters, Nylon, 0.2μm pore, Sterile - FAQs
View additional product information for Nalgene™ Syringe Filters - FAQs (723-2580, 723-9945, 171-0020PK, 723-2520, 723-9920, 171-0045PK, 723-2545, 720-1320, 722-2520, 725-2545, 725-2520, 726-2545, 726-2520, 171-0045, 171-0020)
1 product FAQs found
Are Nalgene Sterile Syringe Filters packed individually or in bulk?
The Nalgene Sterile Syringe Filters are individually blister-packed. The Nalgene non-Sterile Syringe Filters are bulk packed.