BioBasic™ SCX HPLC Columns
Thermo Scientific™

BioBasic™ SCX HPLC Columns

BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
73205-01400110 mm4 mm
73205-05213050 mm2.1 mm
73205-05303050 mm3 mm
73205-101030100 mm1 mm
73205-102130100 mm2.1 mm
73205-103030100 mm3 mm
73205-104630100 mm4.6 mm
73205-151030150 mm1 mm
73205-152130150 mm2.1 mm
73205-154630150 mm4.6 mm
73205-251030250 mm1 mm
73205-254630250 mm4.6 mm
73205-05463050 mm4.6 mm
73205-01300110 mm3 mm
Catalog number 73205-014001
Price (USD)
560.42
Pack of 4
Length (Metric):
10 mm
Diameter (Metric):
4 mm
Price (USD)
560.42
Pack of 4

Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.

  • Strong cation exchange phase based on sulfonic acid chemistry
  • Separation and retention of basic and other cationic species
  • 300Å pore size for enhanced protein and peptide separations
  • Outstanding stability under demanding pH conditions

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatGuard Cartridge, 4 Pk.
Column TypeIon Exchange
EndcappedNo
Max. Pressure5800 psi (400 bar)
Packing MaterialSilica
Stationary PhaseSulfonic Acid
Surface Area100 m²/g
Temperature60°C
USP TypeL52
Carbon Load3%
Diameter (Metric)4 mm
Length (Metric)10 mm
Particle Size5 μm
PhaseIon Exchange
Pore Size300 Å
Product LineBioBasic
TechniqueBio-LC
TypeGuard Cartridge
pH2 to 8
Unit SizePack of 4