Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capacity, holds 320 boxes; 208-230V/60Hz, 12A - FAQs
Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capacity, holds 320 boxes; 208-230V/60Hz, 12A - FAQs
View additional product information for Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (7320D)