This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now

CHROMagar® O157 Medium

Catalog number: 74002

Applied Biosystems™  Related applications: Diagnostic Testing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recommended as a second media to CT-SMAC. CHROMagar® distinguishes between Pseudomonas spp and E.coli O157 by a specific colour reaction.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: Agar Plates
Preparation Method: Microwave
Form: Powder
Target Organism Class: E. coli
Product Line: CHROMagar®
Product Size: 4 x 250 mL
Strain or Genotype: E. coli O157
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products