Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, holds 400 boxes; 115V/60Hz, 16A - FAQs
Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, holds 400 boxes; 115V/60Hz, 16A - FAQs
View additional product information for Forma™ 7000 Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (7400A)