Matrix™ Filtered Pipette Tips, 125 - FAQs
Matrix™ Filtered Pipette Tips, 125 - FAQs
View additional product information for Matrix™ Filtered 384 Format Pipette Tips - FAQs (7445)
1 product FAQs found
Can I install the Complete Stainless Steel Rollout Drawer Set myself?
The Complete Stainless Steel Rollout Drawer Set is a convenient alternative to open-wire shelving and is an option for Thermo Scientific high-performance refrigerators. The product cannot be installed by the user; it needs to factory-installed and this must be specified when ordering the refrigerator.