Search
View additional product information for Complete Stainless Steel Rollout Drawer Set - FAQs (7134, 7445TS, 6701, 6703)
1 product FAQs found
The Complete Stainless Steel Rollout Drawer Set is a convenient alternative to open-wire shelving and is an option for Thermo Scientific high-performance refrigerators. The product cannot be installed by the user; it needs to factory-installed and this must be specified when ordering the refrigerator.