H-Flex 1  Blood Tube Package
H-Flex 1  Blood Tube Package
H-Flex 1  Blood Tube Package
H-Flex 1  Blood Tube Package
H-Flex 1  Blood Tube Package
Thermo Scientific™
H-Flex 1  Blood Tube Package
H-Flex 1 rotor blood tube package offers the flexibility to tun microplates, 5/7 mL and 10/12 mL blood tubes in the same rotor.
Have Questions?
Catalog number 75000156
Price (USD)
-
H-Flex 1 rotor blood processing package containing adapters for microplates, 5/7 ml and 10/12 ml blood tubes
  • H-Flex 1 rotor blood processing package containing adapters for microplates, 5/7 mL and 10/12 mL blood tubes
Specifications
Certifications/ComplianceBuckets are available with biocontainment certified lids
DescriptionBuckets are available with biocontainment certified lids
Includes
  • 75003300 H-Flex 1 Rotor
  • 75003301 H-Flex 1 Buckets
  • 75003302 H-Flex 1 Lids
  • 75003307 Set of 2 Microplate Carriers, 5 standard, 1 deepwell
  • 75003305 Set of 2 adapters, 33 x 10/12 ml blood tubes
  • 75003306 Set of 2 adapters 42 x 5/7 ml blood tubes
Product TypeCan be used with Thermo Scientific Sorvall Legend X1, X1 Pro and ST16, ST1 Plus Series, Thermo Scientific Multifuge X1, X1 Pro and Megafuge 16, ST1 Plus Series, Thermo Scientific SL 16, ST1 Plus Series
Capacity2x10 standard microplates (2x1 deepwell), 66 x 10/12 mL 84 x 5/7 mL blood tubes
Max. RCF4,700 x g
Max. Speed4297
Product LineSorvall
Unit SizeEach