UltraPure™ DEPC-Treated Water
Invitrogen™
UltraPure™ DEPC-Treated Water
Autoclaved to remove the DEPC and 0.1-μm membrane-filtered.
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1.25 mL
1 L
4 x 100 mL
Catalog number 750023
Price (USD)
109.65
Online exclusive
120.00
Save 10.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
109.65
Online exclusive
120.00
Save 10.35
Each
UltraPure™ DEPC-treated Water is suitable for use with RNA. It is prepared by incubating with 0.1% diethylpyrocarbonate (DEPC), and is then autoclaved to remove the DEPC. Sterile filtered.
Performance and Quality Testing: No DNase or RNase activity detected.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineUltraPure
Purification MethodAutoclaved, Membrane-Filtered
Recommended StorageStore at room temperature.
Shipping ConditionRoom Temperature
Treatment(s)DEPC-Treated
GradeMolecular Biology
Quantity1 L
Unit SizeEach
Frequently asked questions (FAQs)
What is the shelf life of UltraPure DEPC-Treated Water?
The UltraPure DEPC-Treated Water is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.