MicroCL 17 Microcentrifuge, ventilated, 230V, with dual row 18 x 2.0/0.5mL rotor - FAQs
MicroCL 17 Microcentrifuge, ventilated, 230V, with dual row 18 x 2.0/0.5mL rotor - FAQs
View additional product information for MicroCL 17 Microcentrifuge - FAQs (75002449)
1 product FAQs found
What rotor does the MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002449) come with?
The MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002449) comes with a Dual Row 18 x 2.0/0.5 mL rotor with screw-on lid.