MicroCL 17 Microcentrifuge, ventilated, 230V, with 24 x 1.5/2.0mL rotor - FAQs
MicroCL 17 Microcentrifuge, ventilated, 230V, with 24 x 1.5/2.0mL rotor - FAQs
View additional product information for MicroCL 17 Microcentrifuge - FAQs (75002450)
1 product FAQs found
What rotor does the MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002450) come with?
The MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002450) comes with a 24 x 1.5/2.0 mL Rotor ClickSeal Biocontainment Lid.