MicroCL 21 Microcentrifuge, ventilated, 120V, with 24 x 1.5/2.0mL rotor - FAQs
MicroCL 21 Microcentrifuge, ventilated, 120V, with 24 x 1.5/2.0mL rotor - FAQs
View additional product information for MicroCL 21 Microcentrifuge - FAQs (75002466)
1 product FAQs found
What rotor does the MicroCL 21 Microcentrifuge (Cat. No. 75002466) come with?
The MicroCL 21 Microcentrifuge (Cat. No. 75002466) comes with a 24 x 1.5/2.0 mL Rotor ClickSeal Biocontainment Lid.