MicroCL 21R Microcentrifuge, refrigerated, 230V, with 24 x 1.5/2.0mL rotor - FAQs
View additional product information for MicroCL 21R Microcentrifuge - FAQs (75002552, 75002561, 75002471, 75002470)
1 product FAQs found
What rotor does the MicroCL 21R Microcentrifuge, Refrigerated (Cat. No. 75002470) come with?
The MicroCL 21R Microcentrifuge (Cat. No. 75002470) comes with a 24 x 1.5/2.0 mL rotor ClickSeal biocontainment lid.