MicroCL 21R Microcentrifuge, refrigerated, 120V, with 24 x 1.5/2.0mL rotor - FAQs
MicroCL 21R Microcentrifuge, refrigerated, 120V, with 24 x 1.5/2.0mL rotor - FAQs
View additional product information for MicroCL 21R Microcentrifuge - FAQs (75002471)
1 product FAQs found
What rotor does the MicroCL 21R Microcentrifuge, Refrigerated (Cat. No. 75002471) come with?
The MicroCL 21R Microcentrifuge (Cat. No. 75002471) comes with a 24 x 1.5/2.0 mL rotor ClickSeal biocontainment lid.