MicroCL 17 Microcentrifuge, ventilated, 230V, with hematocrit rotor - FAQs
MicroCL 17 Microcentrifuge, ventilated, 230V, with hematocrit rotor - FAQs
View additional product information for MicroCL 17 Microcentrifuge - FAQs (75002495)
1 product FAQs found
What rotor does the MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002495) come with?
The MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002495) comes with a Hematocrit rotor.